Learn More
Berkshire™ Open-Cell Foam Lab-Tips™ Swabs
Product Code. 17853232 Shop All Berkshire Products
Click to view available options
Length (Metric):
71 mm
76 mm
127 mm
162 mm
Width (Metric) Tip:
3 mm
3.7 mm
3.8 mm
4 mm
5 mm
13 mm
Unit Size:
Each
Pack of 1000
Pack of 20
Product Code. 17853232
Brand: Berkshire™ LTO1465.10
Only null left
Add to basket
Add to basket
General-purpose swab for cleaning small and confined areas
  • Polyurethane foam over rigid core
  • 100 ppi open-cell foam
  • Thermally bonded head
  • Low particles, fibers, ions and extractables, no contaminating adhesives
  • Good chemical resistance
  • Good sorbency and excellent solvent hold
  • Laundered and packaged in cleanroom
Specifications
Certifications/Compliance ISO Class 4
Diameter (English) Handle 0.12 in.
Length (English) 6.3 in.
For Use With (Application) For cleaning small and confined areas, good chemical resistance, sorbency and excellent solvent hold
Product Type Swab
Diameter (Metric) Handle 3.1 mm
Length (Metric) 162 mm
Length (English) Handle 5.71 in.
Length (Metric) Handle 14.5 mm
Material Polyurethane Foam
Thickness (Metric) 2.8 mm
Length (English) Tip 0.66 in.
Length (Metric) Tip 16.8 mm
Width (Metric) Tip 3.8 mm
Width (English) Tip 0.1 in.
Tip Style Flexible