Learn More
Berkshire™ Open-Cell Foam Lab-Tips™ Swabs
Shop All Berkshire Products
Click to view available options
Length (Metric):
71 mm
76 mm
127 mm
162 mm
Width (Metric) Tip:
3 mm
3.7 mm
3.8 mm
4 mm
5 mm
13 mm
Unit Size:
Each
Pack of 1000
Pack of 20
Description
- Polyurethane foam over rigid core
- 100 ppi open-cell foam
- Thermally bonded head
- Low particles, fibers, ions and extractables, no contaminating adhesives
- Good chemical resistance
- Good sorbency and excellent solvent hold
- Laundered and packaged in cleanroom
Specifications
Specifications
| Certifications/Compliance | ISO Class 4 |
| Length (English) | 2.8 in. |
| For Use With (Application) | Designed for the highest level of contamination control in critical processing applications, cleaning of sensitive equipment, applying lubricants and other liquids, removing excess materials |
| Product Type | Swab |
| Length (Metric) | 71 mm |
| Length (English) Handle | 2.32 in. |
| Length (Metric) Handle | 59 mm |
| Material | Polyurethane Foam |
| Thickness (Metric) | 2 mm |
| Length (English) Tip | 0.43 in. |
| Show More |