missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OPLAH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 540.75
Specifications
| Antigen | OPLAH |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18103229
|
Novus Biologicals
NBP2-47285 |
0.1 mL |
€ 572.00 € 540.75 / 0.10mL Save € 31.25 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632636
|
Novus Biologicals
NBP2-47285-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OPLAH Polyclonal specifically detects OPLAH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| OPLAH | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 26873 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MDPVHIHRHSGLLSALGLALADVVHEAQEPCSLLYAPETFVQLDQRLSRLEEQCVDALQAQGFPRSQISTESFLHLRYQGTDCALMVSAHQH | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 5-Opase, OPLA, OPLAHD | |
| OPLAH | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.