Learn More
OPN1SW Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24660-1-AP-20UL
This item is not returnable.
View return policy
Description
The OPN1SW antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human OPN1SW. This antibody recognizes human, mouse antigen. The OPN1SW antibody has been validated for the following applications: IF, ELISA analysis.
Specifications
| OPN1SW | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Immunofluorescence, ELISA | |
| Blue sensitive opsin, BOP, CBT, OPN1SW | |
| 20 μL | |
| 611 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |