Learn More
OPN1SW Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24660-1-AP-20UL
 View more versions of this product
Product Code. 18077375
  • € 145.00 / 20µL
Description
The OPN1SW antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human OPN1SW. This antibody recognizes human, mouse antigen. The OPN1SW antibody has been validated for the following applications: IF, ELISA analysis.
Specifications
Specifications
OPN1SW
350 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Immunofluorescence, ELISA
Blue sensitive opsin, BOP, CBT, OPN1SW
20 μL
611
Store at -20°C. Stable for one year after shipment.