Learn More
Invitrogen™ Oportuzumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559632
 View more versions of this product
Product Code. 30284100
  • € 1204.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
VB4-845 is studied in the treatment of certain types of head and neck cancer. VB4-845 is made by linking a monoclonal antibody fragment to a toxic protein that may kill cancer cells. VB4-845 is a fusion protein containing humanized scFv specific for the epithelial cell adhesion molecule, Ep-CAM, a tumor cell-associated target highly expressed on carcinoma cells of epithelial origin and a truncated portion of Pseudomonas exotoxin A.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Oportuzumab Humanized
Recombinant Monoclonal
Unconjugated
VB4-845
Protein A
RUO
Human
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized