missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Opsin 5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16528031
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16528031 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16528031 Supplier Invitrogen™ Supplier No. PA590869

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: U-251MG, HeLa, Mouse eye, Mouse brain, Mouse testis, Rat eye, Rat brain; Cellular Location: Membrane, Multi-pass membrane protein.

Opsins are members of the guanine nucleotide-binding protein (G protein)-coupled receptor superfamily. This opsin gene is expressed in the eye, brain, testes, and spinal cord. This gene belongs to the seven-exon subfamily of mammalian opsin genes that includes peropsin (RRH) and retinal G protein coupled receptor (RGR). Like these other seven-exon opsin genes, this gene may encode a protein with photoisomerase activity. Alternative splicing results in multiple transcript variants encoding different isoforms. Detected in the eye, testis, brain, retina and cell lines derived from neural retina.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Opsin 5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene OPN5
Gene Accession No. Q6U736, Q6VZZ7
Gene Alias DJ402H5.1; G protein-coupled receptor 136; Gpr136; G-protein coupled receptor 136; G-protein coupled receptor PGR12; GRP136; Neuropsin; OPN5; opsin 5; opsin-5; Pgr12; TMEM13; Transmembrane protein 13
Gene Symbols OPN5
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221391, 316259, 353344
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.