Learn More
Hellma™ Optical Glass Large Cells
Click to view available options
Capacity (Metric):
20,000 μL
Unit Size:
Each
Specifications
Specifications
| Type | Large Cell |
| Capacity (Metric) | 20,000 μL |
| Material | Optical Glass |
| Width (Metric) Interior | 50mm |
| Quantity | 1 Each |
| Path Length | 10 ±0.2 mm |
| Height (Metric) | 53 mm |
| Width (Metric) | 55 mm |