Learn More
Hellma™ Special Optical Glass Precision Micro Cell Cuvettes
Click to view available options
Capacity (Metric):
35,000 μL
Unit Size:
Each
Description
- Transmission of more than 80% over a spectral range of between 320 and 2500nm
- For photometry and colorimetry measurements
Specifications
Specifications
| Type | Macro Absorption Cell |
| Capacity (Metric) | 35,000 μL |
| Material | Optical Glass |
| Width (Metric) Interior | 9.5mm |
| Path Length | 100 mm |
| Wavelength Range | 360 to 2500 nm |
| Height (Metric) | 45 mm |
| Width (Metric) | 12.5 mm |