Learn More
Macherey-Nagel™ Optima™ 1301 GC Capillary Column
Product Code. 10746142 Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
1 μm
Unit Size:
Each
Product Code. 10746142
Brand: Macherey-Nagel™ 726771.30
Only null left
Specifications
Diameter (Metric) 0.4 mm
Type GC Capillary Column
Diameter (Metric) Inner 0.25 mm
Stationary Phase Optima™ 1301
USP Type G1, G2, G38
For Use With (Application) Pesticide Analysis
Temperature Range 300°C to 320°C
Film Thickness 0.25 μm
Length (Metric) 30 m
Quantity 1 Each