Learn More
Macherey-Nagel™ Optima™ 5 Amine GC Capillary Column
Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
10 m
25 m
30 m
Diameter (Metric) Inner:
0.1 mm
0.2 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.35 μm
0.4 μm
0.5 μm
1 μm
1.5 μm
3 μm
Unit Size:
Each
Spécification
Spécification
| Diameter (Metric) | 0.5 mm |
| Type | GC Capillary Column |
| Diameter (Metric) Inner | 0.32 mm |
| Stationary Phase | Optima™ 5 Amine |
| USP Type | G1, G2, G38 |
| For Use With (Application) | Gas Chromatography (GC), Polyfunctional Amines Analysis |
| Temperature Range | 300°C to 320°C |
| Film Thickness | 1 μm |
| Length (Metric) | 30 m |
| Quantity | 1 Each |