Learn More
Macherey-Nagel™ Optima™ 5 MS GC Capillary Column
Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
12 m
15 m
25 m
30 m
50 m
60 m
Diameter (Metric) Inner:
0.2 mm
0.25 mm
0.32 mm
Film Thickness:
0.2 μm
0.25 μm
0.35 μm
0.5 μm
1 μm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) | 0.4 mm |
| Type | GC Capillary Column |
| Diameter (Metric) Inner | 0.2 mm |
| Stationary Phase | Optima™ 5 MS |
| USP Type | G27, G36 |
| For Use With (Application) | GC/MS, ECD |
| Temperature Range | 320°C to 340°C |
| Film Thickness | 0.2 μm |
| Length (Metric) | 25 m |
| Quantity | 1 Each |