Specifications
| Diameter (Metric) | 0.4 mm |
| Type | Capillary Column |
| Diameter (Metric) Inner | 0.25 mm |
| Stationary Phase | Optima™ BioDiesel F |
| For Use With (Application) | Gas Chromatography (GC) |
| Temperature Range | 240°C to 250°C |
| Length (Metric) | 30 m |
| Quantity | 1 Each |