Learn More
Grant Instruments™ Optima Bridge Plate
View and access thermostat on heating and cooling circulators. Grant Instruments™ Optima Bridge Plate is for use with Grant refrigeration units, heating and cooling circulators.
Brand: Grant Instruments™ T-BPS
Additional Details : Weight : 1.00000kg
Description
View and access thermostat on heating and cooling circulators. Grant Instruments™ Optima Bridge Plate is for use with Grant refrigeration units, heating and cooling circulators.
- For use with Grant refrigeration units, heating and cooling circulators
- Ensures visibility/accessibility of the thermostat whilst optimising bench space
Specifications
T Optima Heating Circulator models to P5 | |
Plastic |