missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR10A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524349
This item is not returnable.
View return policy
Description
OR10A4 Polyclonal Antibody for Western Blot
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
| OR10A4 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9H209 | |
| OR10A4 | |
| KLH conjugated synthetic peptide between 245-273 amino acids from the C-terminal region of human OR10A4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| OR10A4 | |
| HP2; hP2 olfactory receptor; JCG5; olfactory receptor 10A4; olfactory receptor family 10 subfamily A member 4; olfactory receptor OR11-87; olfactory receptor, family 10, subfamily A, member 4 pseudogene; olfactory receptor-like protein JCG5; OR10A4; OR10A4P | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 283297 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction