Learn More
Invitrogen™ OR10G2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5102115
Product Code. 16698584
  • € 493.00 / 100µL
Description
Antibody detects endogenous levels of total OR10G2.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OR10G2
Polyclonal
Unconjugated
OR10G2
olfactory receptor 10G2; olfactory receptor family 10 subfamily G member 2; olfactory receptor OR14-41 pseudogene; olfactory receptor, family 10, subfamily G, member 2; OR10G2; OR14-41
Rabbit
Affinity chromatography
RUO
26534
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8NGC3
OR10G2
A synthesized peptide derived from human OR10G2(Accession Q8NGC3), corresponding to amino acid residues S244-T294.
100 μL
Primary
Human, Mouse, Rat