missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR10P1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139924
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DIVPRTLANL GSLHPQAISF QGCAAP Highest antigen sequence indentity to the following orthologs: Rat - 73%, Mouse - 73%.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
| OR10P1 | |
| Polyclonal | |
| Unconjugated | |
| OR10P1 | |
| Olfactory receptor 10P1; olfactory receptor 10P2; Olfactory receptor 10P3; olfactory receptor family 10 subfamily P member 1; olfactory receptor OR12-7; olfactory receptor, family 10, subfamily P, member 1 pseudogene; olfactory receptor, family 10, subfamily P, member 2 pseudogene; olfactory receptor, family 10, subfamily P, member 3 pseudogene; OR10P1; OR10P1P; OR10P2P; OR10P3P; OR12-7; OST701; seven transmembrane helix receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 121130 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NGE3 | |
| OR10P1 | |
| Recombinant protein corresponding to Human OR10P1. Recombinant protein control fragment (Product #RP-109388). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction