missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR1A2 Polyclonal Antibody

OR1A2 Polyclonal Antibody
Brand: Invitrogen PA570835
This item is not returnable.
View return policy
Description
OR1A2 Polyclonal Antibody for Western Blot
Specifications
| OR1A2 | |
| Polyclonal | |
| Unconjugated | |
| OR1A2 | |
| Olfactory receptor 17-6; olfactory receptor 1A2; olfactory receptor family 1 subfamily A member 2; olfactory receptor OR17-10; olfactory receptor, family 1, subfamily A, member 2; OR17-6; OR1A2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26189 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y585 | |
| OR1A2 | |
| synthetic peptide directed towards the C-terminal region of human OR1A2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction