missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR2A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534047
This item is not returnable.
View return policy
Description
OR2A4 Polyclonal Antibody for IHC (P)
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
| OR2A4 | |
| Polyclonal | |
| Unconjugated | |
| OR2A4 | |
| Olfactory receptor 2A10; olfactory receptor 2A4; olfactory receptor family 2 subfamily A member 4; olfactory receptor OR6-37; olfactory receptor, family 2, subfamily A, member 10; olfactory receptor, family 2, subfamily A, member 4; OR2A10; OR2A4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79541 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O95047 | |
| OR2A4 | |
| Synthetic 18 amino acid peptide from 3rd cytoplasmic domain of human OR2A4. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction