Learn More
OR2T12 Polyclonal Antibody
OR2T12 Polyclonal Antibody
Brand: Invitrogen PA571176
This item is not returnable.
View return policy
Description
OR2T12 Polyclonal Antibody for Western Blot
Specifications
| OR2T12 | |
| Polyclonal | |
| Unconjugated | |
| OR2T12 | |
| novel 7 transmembrane receptor (rhodopsin family) protein; olfactory receptor 2T12; olfactory receptor family 2 subfamily T member 12; olfactory receptor OR1-57; olfactory receptor, family 2, subfamily T, member 12; OR1-57; OR2T12 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 127064 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NG77 | |
| OR2T12 | |
| synthetic peptide directed towards the C-terminal region of human OR2T12. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |