missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR2Z1 Polyclonal Antibody

Product Code. 15739621 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15739621 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15739621 Supplier Invitrogen™ Supplier No. PA561814

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MASDFLRGEG ATSYGGGAAQ I Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 57%.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2Z1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OR2Z1
Gene Accession No. Q8NG97
Gene Alias Olfactory receptor 2Z1; olfactory receptor 2Z2; olfactory receptor family 2 subfamily Z member 1; olfactory receptor OR19-4; olfactory receptor, family 2, subfamily Z, member 1; olfactory receptor, family 2, subfamily Z, member 2; OR19-4; OR2Z1; OR2Z2
Gene Symbols OR2Z1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OR2Z1. Recombinant protein control fragment (Product #RP-100880).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284383
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.