missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR4A4/OR4A47 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538228
This item is not returnable.
View return policy
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
| OR4A4/OR4A47 | |
| Polyclonal | |
| Unconjugated | |
| OR4A4P | |
| OR4A47, OR4A4P | |
| A synthetic peptide derived from the C-terminal region of human OR4A4/4A47 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6IF82, Q8NGN8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 390134, 403253 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction