missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR4F17 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17147468 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17147468 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17147468 Supplier Invitrogen™ Supplier No. PA5116232

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OR4F17 Polyclonal Antibody for Western Blot

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR4F17
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR4F17
Gene Accession No. Q8NGA8
Gene Alias Olfactory receptor 4F11; olfactory receptor 4F17; Olfactory receptor 4F18; olfactory receptor 4F19; olfactory receptor family 4 subfamily F member 17; olfactory receptor, family 4, subfamily F, member 11 pseudogene; olfactory receptor, family 4, subfamily F, member 17; olfactory receptor, family 4, subfamily F, member 19; OR4F11P; OR4F17; OR4F18; OR4F19
Gene Symbols OR4F17
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 61-89 amino acids from the N-terminal region of human OR4F17.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81099
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.