Learn More
OR4K1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-05843MFV450
Product Code. 18470599
  • € 428.00 / 0.10mL
Description
OR4K1 Polyclonal antibody specifically detects OR4K1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50mM Sodium Borate
Produced in rabbits immunized with E. coli-derived Human OR4K1 fragment.