missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR51G1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16659684
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16659684 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16659684 Supplier Invitrogen™ Supplier No. PA5103233

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR51G1.

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR51G1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR51G1
Gene Accession No. Q8NGK1
Gene Alias Olfactory receptor 51G1; olfactory receptor 51G3; olfactory receptor family 51 subfamily G member 1 (gene/pseudogene); olfactory receptor OR11-29; olfactory receptor, family 51, subfamily G, member 1; OR11-29; OR51G1; OR51G3P
Gene Symbols OR51G1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR51G1(Accession Q8NGK1), corresponding to amino acid residues C207-M257.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79324
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.