missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OR51L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548351
This item is not returnable.
View return policy
Description
OR51L1 Polyclonal Antibody for Western Blot
Odorant receptor.
Specifications
| OR51L1 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8NGJ5 | |
| OR51L1 | |
| KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human OR51L1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| OR51L1 | |
| Olfactory receptor 51L1; olfactory receptor family 51 subfamily L member 1; olfactory receptor OR11-31; olfactory receptor, family 51, subfamily L, member 1; OR11-31; OR51L1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 119682 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction