missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR52I2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06488-50ul
This item is not returnable.
View return policy
Description
OR52I2 Polyclonal antibody specifically detects OR52I2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| OR52I2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| olfactory receptor 52I2, olfactory receptor, family 52, subfamily I, member 2, OR11-12 | |
| Produced in rabbits immunized with E. coli-derived Human OR52I2 fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 143502 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction