Learn More
Invitrogen™ OR56A3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA571231
Product Code. 15819643
  • € 520.00 / 100µL
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 100%.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OR56A3
Polyclonal
Unconjugated
OR56A3
Olfactory receptor 56A3; olfactory receptor 56A6; olfactory receptor family 56 subfamily A member 3; olfactory receptor OR11-51; olfactory receptor, family 56, subfamily A, member 2 pseudogene; olfactory receptor, family 56, subfamily A, member 3 pseudogene; olfactory receptor, family 56, subfamily A, member 6; OR56A2P; OR56A3; OR56A3P; OR56A6
Rabbit
Affinity Chromatography
RUO
390083
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8NH54
OR56A3
synthetic peptide directed towards the C-terminal region of human OR56A3.