missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR6B3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17290194 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17290194 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17290194 Supplier Invitrogen™ Supplier No. PA5117700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR6B3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene OR6B3
Gene Accession No. Q8NGW1
Gene Alias olfactory receptor 6B3; olfactory receptor family 6 subfamily B member 3; olfactory receptor OR2-2; olfactory receptor, family 6, subfamily B, member 3 pseudogene; OR6B3; OR6B3P; OR6B3Q
Gene Symbols OR6B3
Host Species Rabbit
Immunogen Peptide sequence from Human Olfactory receptor 6B3 protein (259-272aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150681
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.