missing translation for 'onlineSavingsMsg'
Learn More

OR6S1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16668694
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16668694 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16668694 Supplier Invitrogen Supplier No. PA5102222

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

OR6S1 Polyclonal Antibody

OR6S1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen OR6S1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR6S1
Gene Accession No. Q8NH40
Gene Alias Olfactory receptor 6S1; olfactory receptor family 6 subfamily S member 1; olfactory receptor OR14-37; olfactory receptor, family 6, subfamily S, member 1; OR14-37; OR6S1; OR6S1Q
Gene Symbols OR6S1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR6S1(Accession Q8NH40), corresponding to amino acid residues Q173-L223.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 341799
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.