Learn More
OR6T1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99229MFV450
Product Code. 18544202
  • € 428.00 / 0.10mL
Description
OR6T1 Polyclonal antibody specifically detects OR6T1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot
olfactory receptor 6T1, Olfactory receptor OR11-277, olfactory receptor, family 6, subfamily T, member 1, OR11-277
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human OR6T1.