missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OR7C1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15759251 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15759251 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15759251 Supplier Invitrogen™ Supplier No. PA561436

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ETGNQTHAQE FLLLGFSATS EIQF Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 58%.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR7C1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OR7C1
Gene Accession No. O76099
Gene Alias CIT-HSP-146E8; HSTPCR86P; olfactory receptor 7C1; Olfactory receptor 7C4; olfactory receptor family 7 subfamily C member 1; olfactory receptor OR19-16; Olfactory receptor TPCR86; olfactory receptor, family 7, subfamily C, member 1; olfactory receptor, family 7, subfamily C, member 4; OR19-5; OR7C1; OR7C4; TPCR86
Gene Symbols OR7C1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OR7C1. Recombinant protein control fragment (Product #RP-100789).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26664
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.