missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orai1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77289AF750
This item is not returnable.
View return policy
Description
Orai1 Polyclonal antibody specifically detects Orai1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Orai1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| CRACM1ORAT1, FLJ14466, ORAI calcium release-activated calcium modulator 1, Protein orai-1, TMEM142Acalcium release-activated calcium modulator 1, Transmembrane protein 142Acalcium release-activated calcium channel protein 1 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human ORAI1. The immunogen is located within the last 50 amino acids of ORAI1. Amino Acid Squence: QDQLDHRGDHPLTPG | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 84876 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction