missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orai1 Antibody (Amgen patent anti-ORAI1), Janelia Fluor™ 646, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27962JF646
This item is not returnable.
View return policy
Description
Orai1 Monoclonal antibody specifically detects Orai1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Orai1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| CRACM1ORAT1, FLJ14466, ORAI calcium release-activated calcium modulator 1, Protein orai-1, TMEM142Acalcium release-activated calcium modulator 1, Transmembrane protein 142Acalcium release-activated calcium channel protein 1 | |
| Orai1 | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 84876 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| Amgen patent anti-ORAI1 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction