Learn More
ORAI1 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28411-1-AP-150UL
 View more versions of this product
Product Code. 17221406
  • € 410.00 / 150µL
Description
Store-operated calcium channels (SOCs) play essential functions in various physiological processes. ORAI1 is a complex glycosylated protein, it exists in several molecular weight forms: a theoretical molecular weight of 32.7 kDa of the unmodified protein and two major glycosylated proteins of ∼43 kDa and ∼50 kDa (PMID: 22318387). Human patients with a severe combined immunodeficiency (SCID) syndrome possess point mutations in Orai1 gene.
Specifications
Specifications
ORAI1
Polyclonal
Unconjugated
150 μL
84876
Liquid
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
300 μg/mL