Learn More
ORAI1 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28411-1-AP-20UL
Description
Store-operated calcium channels (SOCs) play essential functions in various physiological processes. ORAI1 is a complex glycosylated protein, it exists in several molecular weight forms: a theoretical molecular weight of 32.7 kDa of the unmodified protein and two major glycosylated proteins of ∼43 kDa and ∼50 kDa (PMID: 22318387). Human patients with a severe combined immunodeficiency (SCID) syndrome possess point mutations in Orai1 gene.
Specifications
| ORAI1 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 84876 | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |