missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORAI3 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98004DL594
This item is not returnable.
View return policy
Description
ORAI3 Polyclonal specifically detects ORAI3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| ORAI3 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| ORAI calcium release-activated calcium modulator 3, TMEM142C | |
| Produced in rabbits immunized with E. coli-derived Human ORAI3 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 93129 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction