missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORAI3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82013MFV450
This item is not returnable.
View return policy
Description
ORAI3 Polyclonal antibody specifically detects ORAI3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
Specifications
| ORAI3 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunocytochemistry | |
| ORAI calcium release-activated calcium modulator 3, TMEM142C | |
| ORAI3 antibody was raised against a 15 amino acid synthetic peptide from near the amino terminus of human ORAI3. The immunogen is located within the first 50 amino acids of ORAI3. Amino Acid Squence: DAGEQAPLNPEGESPAGSA | |
| 0.1 mL | |
| Immunology | |
| 93129 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction