Learn More
ORAI3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82014MFV450
This item is not returnable.
View return policy
Description
ORAI3 Polyclonal antibody specifically detects ORAI3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| ORAI3 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| ORAI3 antibody was raised against a 19 amino acid synthetic peptide from near the carboxy terminus of human ORAI3. The immunogen is located within amino acids 200 - 250 of ORAI3. Amino Acid Squence: ASNLPRSSASAAPSQAEPA |
| Western Blot, ELISA, Immunocytochemistry | |
| Rabbit | |
| IgG |