missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ORAI3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA582518

Product Code. 16309484

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: APLDTPTPMV PTSRVPGTLA PVATSLSPAS NLPRSSASAA PSQAEPACPP RQACGGGGAH GPGWQA Highest antigen sequence indentity to the following orthologs: Rat - 63%, Mouse - 58%.

ORAI3 is a protein coding gene that is involved in calcium signaling and regulation of calcium homeostasis. It is a member of the ORAI family of proteins that form calcium release-activated calcium (CRAC) channels in the plasma membrane of cells. These channels are activated by depletion of calcium from the endoplasmic reticulum, leading to an influx of calcium ions into the cytoplasm. ORAI3 has been implicated in a variety of physiological processes, including T-cell activation, muscle contraction, and neurotransmitter release. Dysregulation of ORAI3 expression or function has been associated with various diseases, including immune disorders, cancer, and cardiovascular disease.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ORAI3
Polyclonal
Unconjugated
ORAI3
9930124N15; BC061259; calcium release-activated calcium channel 3; calcium release-activated calcium modulator 3 protein; ORAI calcium release-activated calcium modulator 3; ORAI3; Protein orai-3; RGD1306538; Tmem142c; Transmembrane protein 142C
Rabbit
Antigen affinity chromatography
RUO
93129
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.4 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BRQ5
ORAI3
Recombinant protein corresponding to Human ORAI3. Recombinant protein control fragment (Product #RP-91115).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.