Learn More
Scientific Industries™ Orbital-Genie Shaker Ratcheting Clamps
Product Code. 11351484 Shop All Ohaus Products
Change view
Click to view available options
Speed Range:
10 to 300 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Unit Size
11351484 10 to 300 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11351484 Supplier Scientific Industries™ Supplier No. SI1502
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Powerful orbital shaker for large volumes. No additional accessories necessary.
Large footprint orbital shaker with universal ratcheting clamps provides the flexibility for using a mix of different size and shape vessels on a single platform. Unit is supplied with two ratcheting and one non-ratcheting clamps.
TRUSTED_SUSTAINABILITY
Specifications
Orbit 35 mm
Load Bearing Capacity (Metric) 12 kg
Speed Range 10 to 300 rpm
Voltage 230 V
Model Genie Temp-Shaker 300 with Adhering Mat