missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORC1 Antibody (ORC1 7F6/1), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-121R
This item is not returnable.
View return policy
Description
ORC1 Monoclonal specifically detects ORC1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| ORC1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ORC1 | |
| Bacterially expressed Human ORC1 | |
| 0.1 mL | |
| Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4998 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry | |
| ORC1 7F6/1 | |
| Western Blot | |
| HS, HSORC1, ORC1Lorigin recognition complex subunit 1, origin recognition complex, subunit 1, origin recognition complex, subunit 1 (yeast homolog)-like, origin recognition complex, subunit 1 homolog, origin recognition complex, subunit 1 homolog (S. cerevisiae), origin recognition complex, subunit 1-like, origin recognition complex, subunit 1-like (yeast), PARC1origin recognition complex 1, Replication control protein 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction