missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111111
This item is not returnable.
View return policy
Description
Immunogen sequence: NNVTPYVVSL QAKDCPDMKH FLQKLISQLM DCCVDIKSKE EESVHVTQRK THYSMDSLSS WYMTVTQKTD PKMLSKKRTT SSQWQSPPVV VILKD Highest antigen sequence indentity to the following orthologs: Rat - 76%, Mouse - 71%.
Histone deacetylase 2 (HDAC2), or transcriptional regulator homolog RPD3 L1, is highly homologous to the yeast transcription factor RPD3 (reduced potassium dependency 3) gene. As in yeast, human HDA2 is likely to be involved in regulating chromatin structure during transcription. It has been implicated to associate with YY1, a mammalian zinc-finger transcription factor, which negatively regulates transcription by tethering RPD3 to DNA as a cofactor. This process is highly concerved from yeast to human.
Specifications
| ORC3 | |
| Polyclonal | |
| Unconjugated | |
| ORC3 | |
| homolog of latheo, Drosophila; IMAGE50150; LAT; LATHEO; Orc3; ORC3L; origin of replication 3 homolog; origin recognition complex subunit 3; Origin recognition complex subunit Latheo; origin recognition complex, subunit 3; origin recognition complex, subunit 3 honolog; origin recognition complex, subunit 3-like; OTTHUMP00000040611; RP3-486L4.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23595 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBD5 | |
| ORC3 | |
| Recombinant Protein corresonding to Human ORC3. Recombinant protein control fragment (Product #RP-94839). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction