missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ORC4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17258473 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17258473 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17258473 Supplier Invitrogen™ Supplier No. PA5115693

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ORC4L.

The origin recognition complex (ORC) is a highly conserved six subunit protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. This gene encodes a subunit of the ORC complex. Several alternatively spliced transcript variants, some of which encode the same protein, have been reported for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORC4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ORC4
Gene Accession No. O43929, O88708, Q6P9Z8
Gene Alias HSORC4; mMmORC4; Orc4; ORC4L; Orc4P; Origin recognition complex subunit 4; origin recognition complex, subunit 4; origin recognition complex, subunit 4 homolog; origin recognition complex, subunit 4-like
Gene Symbols ORC4
Host Species Rabbit
Immunogen A synthesized peptide derived from human ORC4(Accession O43929), corresponding to amino acid residues L404-L436.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26428, 295596, 5000
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.