missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ORC6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576314
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The origin recognition complex (ORC) is a highly conserved six subunit protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. Gene silencing studies with small interfering RNA demonstrated that this protein plays an essential role in coordinating chromosome replication and segregation with cytokinesis.
Specifications
| ORC6 | |
| Polyclonal | |
| Unconjugated | |
| Orc6 | |
| 6720420I10Rik; ORC 6; ORC6; ORC6L; origin recognition complex subunit 6; origin recognition complex, subunit 6; origin recognition complex, subunit 6-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23594, 291927, 56452 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9WUJ8, Q9Y5N6 | |
| Orc6 | |
| Recombinant protein of human ORC6. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction