missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORC6L Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74619JF549
This item is not returnable.
View return policy
Description
ORC6L Polyclonal antibody specifically detects ORC6L in Human samples. It is validated for Western Blot
Specifications
| ORC6L | |
| Polyclonal | |
| Western Blot | |
| ORC6Lorigin recognition complex, subunit 6 homolog-like (yeast), origin recognition complex subunit 6, origin recognition complex, subunit 6, origin recognition complex, subunit 6 like (yeast) | |
| The immunogen recognized by this antibody maps to a region between residue 200 and 230 of human origin recognition complex subunit 6 using the numbering given in entry NP_055136.1 (GeneID 23594). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 23594 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction