missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORC6L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP3-38152-100ul
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ORC6L Polyclonal antibody specifically detects ORC6L in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Especificaciones
| ORC6L | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| ORC6Lorigin recognition complex, subunit 6 homolog-like (yeast), origin recognition complex subunit 6, origin recognition complex, subunit 6, origin recognition complex, subunit 6 like (yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-252 of human ORC6L (NP_055136.1).,, Sequence:, MGSELIGRLAPRLGLAEPDMLRKAEEYLRLSRVKCVGLSARTTETSSAVMCLDLAASWMKCPLDRAYLIKLSGLNKETYQSCLKSFECLLGLNSNIGIRDLAVQFSCIEAVNMASKILKSYESSLPQTQQVDLDLSRPLFTSAALLSACKILKLKVDKNKMVATSGVKKAIFDRLCKQLEKIGQQVDREPGDVATPPRKRKKIVVEAPAKEMEKVEEMPHKPQKDEDLTQDYEEWKRKILENAASAQKATAE | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 23594 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido