missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orexin Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 470.00
Specifications
| Antigen | Orexin |
|---|---|
| Dilution | Western Blot 1:100 - 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635890
|
Novus Biologicals
NBP2-93319-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18696981
|
Novus Biologicals
NBP2-93319-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Orexin Polyclonal antibody specifically detects Orexin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Orexin | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Neuronal Cell Markers, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 3060 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| hypocretin (orexin) neuropeptide precursor, NRCLP1, orexin, OXHcrt, PPORX, PPOXHypocretin | |
| A synthetic peptide corresponding to a sequence within amino acids 34-66 of human Orexin (NP_001515.1). QPLPDCCRQKTCSCRLYELLHGAGNHAAGILTL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title