missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orexin R1/HCRTR1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95106-0.1ml
This item is not returnable.
View return policy
Description
Orexin R1/HCRTR1 Polyclonal antibody specifically detects Orexin R1/HCRTR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Orexin R1/HCRTR1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| hypocretin (orexin) receptor 1, hypocretin receptor 1, Hypocretin receptor type 1, hypocretin receptor-1, orexin receptor 1, orexin receptor type 1, orexin receptor-1, ox-1-R, Ox1-R, OX1R | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human HCRTR1 (NP_001516.2). HPLLFKSTARRARGSILGIWAVSLAIMVPQAAVMECSSVLPELANRTRLFSVCDERWADDLYPKIYHSCFFIVTYLAPLGLMAMAYFQIFRKLWGRQIPGT | |
| 0.1 mL | |
| Cancer, GPCR, Neuronal Cell Markers, Neuroscience | |
| 3061 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction