missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orexin R2/HCRTR2 Antibody (456738), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5246V
This item is not returnable.
View return policy
Description
Orexin R2/HCRTR2 Monoclonal antibody specifically detects Orexin R2/HCRTR2 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Orexin R2/HCRTR2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 456738 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| hypocretin (orexin) receptor 2, Hypocretin receptor type 2, orexin receptor type 2, Ox-2-R, Ox2-R, OX2R | |
| NS0 mouse myeloma cell line transfected with human Orexin R2/HCRTR2, Met1-Trp444, Accession # AAC39602 | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, GPCR, Immunology, Neuronal Cell Markers, Neuroscience | |
| 3062 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction