missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Orexin Receptor 2 Polyclonal Antibody

Description
This antibody is predicted to react with bovine, canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein binds the hypothalamic neuropeptides orexin A and orexin B. A related gene encodes a G-protein coupled receptor that selectively binds orexin A.
Specifications
Specifications
| Antigen | Orexin Receptor 2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | HCRTR2 |
| Gene Accession No. | O43614 |
| Gene Alias | HCRTR1; Hcrtr2; hypocretin (orexin) receptor 2; hypocretin receptor 2; hypocretin receptor type 2; mOX2aR; mOX2bR; Mox2r; mOXR2; orexin receptor 1; orexin receptor 2; orexin receptor type 2; orexin receptor-1; orexin receptor-2; orexin type-2 receptor; OTTHUMP00000039969; OX1R; ox2r; ox-2-R; ox2-R |
| Gene Symbols | HCRTR2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?